missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SULT1A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102435
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SULT1A1.
The soluble sulfotransferases contribute to the elimination of xenobiotics, the activation of procarcinogens and the regulation of hormones by catalyzing the sulfate conjugation of these substances. Members of the three groups comprising this superfamily (namely SULT1, SULT2 and SULT3) show selectivity to certain substrate compounds. SULT1A1 (sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1), also known as STP or STP1, is a 295 amino acid protein that localizes to the cytoplasm and belongs to the sulfotransferase family. Functioning as a homodimer that is expressed in brain, liver, skin and lung tissue, SULT1A1 catalyzes the sulfate conjugation of catecholamines, phenolic drugs and neurotransmitters and, via this catalytic activity, plays a role in the elimination of a variety of compounds from the body. Additionally, SULT1A1 may be involved in the activation of carcinogenic N-hydroxyarylamines, indicating a possible role in carcinogenesis.
Specifications
| SULT1A1 | |
| Polyclonal | |
| Unconjugated | |
| SULT1A1 | |
| AI266890; aryl sulfotransferase; Aryl sulfotransferase 1; Aryl sulfotransferase cytosolic 1A phenol-preferring member 3; Aryl sulfotransferase cytosolic, 1A, phenol-preferring, member 3; aryl sulfotransferase IV; arylsulfotransferase ST1A4; ASTIV; CSPS; HAST1/HAST2; MGC131921; MGC5163; Minoxidil sulfotransferase; mSTp1; Mx-ST; OK/SW-cl.88; OTTHUMP00000162568; OTTHUMP00000162569; P PST; P PST 1; Phenol sulfotransferase; Phenol sulfotransferase 1; Phenol sulfotransferase 1a1; phenol/aryl sulfotransferase; phenol-sulfating phenol sulfotransferase 1; P-PST; P-PST 1; PST; PST-1; St1a1; ST1A3; ST1A4; Stm; STP; STP1; sulfokinase; Sulfotransferase 1A1; sulfotransferase family 1A member 1; sulfotransferase family 1A, phenol-preferring, member 1; sulfotransferase family, cytosolic, 1A, phenol preferring; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1; sulfotransferase, phenol preferring 1; sulphotransferase family cytosolic 1A phenol-prefering member 1; SULT1A1; Sult1a3; Thermostable phenol sulfotransferase; thermostable phenol sulfotransferase1; ts-PST; TSPST1; Tyrosine-ester sulfotransferase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20887, 6817, 83783 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P17988, P50225, P52840 | |
| SULT1A1 | |
| A synthesized peptide derived from human SULT1A1(Accession P50225), corresponding to amino acid residues G65-P115. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction