missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SULT1A1 Recombinant Rabbit Monoclonal Antibody (034)
GREENER_CHOICE

Product Code. 16353624
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16353624 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16353624 Supplier Invitrogen™ Supplier No. MA529590

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human SULT1A1.

The soluble sulfotransferases contribute to the elimination of xenobiotics, the activation of procarcinogens and the regulation of hormones by catalyzing the sulfate conjugation of these substances. Members of the three groups comprising this superfamily (namely SULT1, SULT2 and SULT3) show selectivity to certain substrate compounds. SULT1A1 (sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1), also known as STP or STP1, is a 295 amino acid protein that localizes to the cytoplasm and belongs to the sulfotransferase family. Functioning as a homodimer that is expressed in brain, liver, skin and lung tissue, SULT1A1 catalyzes the sulfate conjugation of catecholamines, phenolic drugs and neurotransmitters and, via this catalytic activity, plays a role in the elimination of a variety of compounds from the body. Additionally, SULT1A1 may be involved in the activation of carcinogenic N-hydroxyarylamines, indicating a possible role in carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SULT1A1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Recombinant Monoclonal
Clone 034
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SULT1A1
Gene Accession No. P50225
Gene Alias AI266890; aryl sulfotransferase; Aryl sulfotransferase 1; Aryl sulfotransferase cytosolic 1A phenol-preferring member 3; Aryl sulfotransferase cytosolic, 1A, phenol-preferring, member 3; aryl sulfotransferase IV; arylsulfotransferase ST1A4; ASTIV; CSPS; HAST1/HAST2; MGC131921; MGC5163; Minoxidil sulfotransferase; mSTp1; Mx-ST; OK/SW-cl.88; OTTHUMP00000162568; OTTHUMP00000162569; P PST; P PST 1; Phenol sulfotransferase; Phenol sulfotransferase 1; Phenol sulfotransferase 1a1; phenol/aryl sulfotransferase; phenol-sulfating phenol sulfotransferase 1; P-PST; P-PST 1; PST; PST-1; St1a1; ST1A3; ST1A4; Stm; STP; STP1; sulfokinase; Sulfotransferase 1A1; sulfotransferase family 1A member 1; sulfotransferase family 1A, phenol-preferring, member 1; sulfotransferase family, cytosolic, 1A, phenol preferring; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1; sulfotransferase, phenol preferring 1; sulphotransferase family cytosolic 1A phenol-prefering member 1; SULT1A1; Sult1a3; Thermostable phenol sulfotransferase; thermostable phenol sulfotransferase1; ts-PST; TSPST1; Tyrosine-ester sulfotransferase
Gene Symbols SULT1A1
Host Species Rabbit
Immunogen Recombinant Human SULT1A1 protein (Glu2-Leu295).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6817
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.