Learn More
SULT1A4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11376-1-AP-20UL
 View more versions of this product
Product Code. 16828413
  • € 145.00 / 20µL
Specifications
SULT1A4
Polyclonal
Unconjugated
SULT1A4
Aryl sulfotransferase 1A3/1A4, HAST3, M PST, ST1A3/ST1A4, STM, Sulfotransferase 1A3/1A4, SULT1A3, SULT1A4, TL PST
Rabbit
Antigen Affinity Chromatography
RUO
445329
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
0.45 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P0DMN0
SULT1A4
SULT1A4 Fusion Protein Ag1935
20 μL
Primary
Mouse, Human