missing translation for 'onlineSavingsMsg'
Learn More

SULT1C2 Monoclonal Antibody (OTI5A11), TrueMAB™, OriGene

Product Code. 17567481
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17567481 30 μL 30µL
17547481 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17567481 Supplier Origene Technologies Supplier No. TA502666S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SULT1C2 Monoclonal antibody specifically detects SULT1C2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SULT1C2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5A11
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SULT1C2
Gene Accession No. O00338
Gene Alias (PST)G; 1810008N17Rik; 1C member 4; HAST-I; humSULTC2; LOC100729651; MGC149521; MGC34422; mOLFST; N-hydroxyarylamine sulfotransferase; N-hydroxylarylamine sulfotransferase; phenol sulfotransferase; Phenol sulfotransferase 1c1; P-SULT; rabSULT1C2; rSULT1C2; St1c1; ST1C2; ST1C4; Stp2; sulfotransferase; sulfotransferase 1C1; Sulfotransferase 1C2; sulfotransferase 1C4; sulfotransferase family 1A phenol-preferring member 2; sulfotransferase family 1A, member 2; sulfotransferase family 1A, phenol-preferring, member 2; sulfotransferase family 1C member 2; sulfotransferase family 1C member 4; sulfotransferase family, cytosolic, 1C, member 1; sulfotransferase family, cytosolic, 1C, member 2; sulfotransferase family, cytosolic, 1C, member 3; sulfotransferase family, cytosolic, 1C, member 4; sulfotransferase family, cytosolic, 1C, member C2; sulfotransferase K1; sulfotransferase, phenol preferring 2; sulphotransferase family cytosolic 1A phenol-preferring member 2; Sult1a2; SULT1C; SULT1C#1; SULT1C#2; SULT1C1; SULT1C2; Sult1c3; SULT1C4; Sultk1
Host Species Mouse
Immunogen Full length human recombinant protein of human SULT1C2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6819
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.