Learn More
SULT1E1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12522-1-AP-150UL
 View more versions of this product
Product Code. 16820274
  • € 410.00 / 150µL
Description
This antibody detected a major band around 35 kDa in lysates of mouse kidney and human liver.
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes a protein that transfers a sulfo moiety to and from estrone, which may control levels of estrogen receptors.
Specifications
Specifications
SULT1E1
Polyclonal
Unconjugated
SULT1E1
EST, EST 1, Estrogen sulfotransferase, ST1E1, STE, Sulfotransferase 1E1, SULT1E1
Rabbit
Antigen Affinity Chromatography
RUO
20860, 360268, 6783
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.38 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P49888, P49891, P52844
Sult1e1
SULT1E1 Fusion Protein Ag3216
150 μL
Primary
Human, Mouse, Rat