missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULT2A1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16348-1-AP-150UL
This item is not returnable.
View return policy
Description
SULT1 sulfotransferases exhibit N-sulfating activities of carcinogenic heterocyclic amines, and are selective toward phenols, whereas SULT2 enzymes prefer hydroxysteroids and SULT3 family members are selective for N-substituted aryl and alicyclic compounds. SULT2A1 catalyzes the sulfonation of procarcinogen xenobiotics, hydroxysteroids and bile acids, and is highly expressed in adrenal and liver tissues. SULT2A1 plays a role in hepatic cholesterol homeostasis. SULT2B1 consists of two isoforms, SULT2B1a and SULT2B1b, which are transcribed from the same gene by alternative splicing of their first exons. Both isoforms are highly selective for the sulphation of 3b-hydroxysteroids such as pregnenolone, epiandrosterone, DHEA and androstenediol. SULT2B1b is expressed in prostate, skin, placenta and lung.Specifications
| SULT2A1 | |
| Polyclonal | |
| Unconjugated | |
| SULT2A1 | |
| Bile salt sulfotransferase, DHEA ST, DHEAS, HST, hSTa, ST2, ST2A1, ST2A3, STD, Sulfotransferase 2A1, SULT2A1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6822 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q06520 | |
| SULT2A1 | |
| SULT2A1 Fusion Protein Ag9542 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction