Learn More
Invitrogen™ SULT2B1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140284
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: KAKVIYMGRN PRDVVVSLYH YSKIAGQLKD PGTPDQFLRD FLKGEVQFGS WFDH Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 93%.
Cytosolic sulfotransferases (STs or SULTs) catalyze the sulfate conjugation of many drugs, xenobiotic compounds, hormones, and neurotransmitters. Her et al. (1998) identified an EST with homology to ST enzymes. By PCR with human placental and prostate cDNA as template, they isolated 2 alternatively spliced cDNAs, identical throughout most of their sequences, but having different 5-prime ends. The shorter cDNA, SULT2B1a, encodes a protein of 350 amino acids; the longer cDNA, SULT2B1b, encodes a 365-amino acid protein. Genomic PCR analysis revealed that the gene encoding both cDNAs, SULT2B1, contains 7 exons, with 2 alternative first exons being used to generate SULT2B1a and SULT2B1b. Northern blot analysis revealed that the SULT2B1 gene is expressed as a 1.4-kb transcript predominantly in prostate, placenta, and trachea.
Specifications
| SULT2B1 | |
| Polyclonal | |
| Unconjugated | |
| SULT2B1 | |
| AI326997; alcohol sulfotransferase; BB173635; HSST2; Hydroxysteroid sulfotransferase 2; RGD:1308882}; ST2B1; sulfotransferase 2B; sulfotransferase 2B1; Sulfotransferase family 2B member 1; sulfotransferase family cytosolic 2B member 1; sulfotransferase family, cytosolic, 2B, member 1; SULT2B; Sult2b1; sult2b1 {ECO:0000312; SULT2B1a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6820 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00204 | |
| SULT2B1 | |
| Recombinant protein corresponding to Human SULT2B1. Recombinant protein control fragment (Product #RP-109558). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.