missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SULT6B1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569543
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 92%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 92%; Rat: 86%.
SULT6B1 is a protein coding gene.
Specifications
| SULT6B1 | |
| Polyclonal | |
| Unconjugated | |
| SULT6B1 | |
| 2410078J06Rik; AV101767; ST6B1; Sulfotransferase 6B1; sulfotransferase family 6B member 1; sulfotransferase family, cytosolic, 6B, member 1; sulfotransferase SULT6B1; SULT6B1; Thyroxine sulfotransferase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 391365 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6IMI4 | |
| SULT6B1 | |
| synthetic peptide directed towards the C terminal region of human SULT6B1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction