Learn More
SUMO1 Antibody (SPM571), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34805MFV610
This item is not returnable.
View return policy
Description
SUMO1 Monoclonal antibody specifically detects SUMO1 in Human samples. It is validated for Flow Cytometry
Specifications
| SUMO1 | |
| SPM571 | |
| Flow Cytometry | |
| 0.1 mL | |
| Purified |
| Flow Cytometry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Alzheimers Research, Core ESC Like Genes, Neuroscience, Stem Cell Markers |