missing translation for 'onlineSavingsMsg'
Learn More

SUMO1 Mouse anti-Human, Rat, Clone: SUMO1/1188, Abnova™

Product Code. 16062936
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16062936 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16062936 Supplier Abnova Supplier No. MAB14625.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant human SUMO1.

  • High specificity
  • High reactivity

Specifications

Antigen SUMO1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SUMO1/1188
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human SUMO1.
Dilution Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined
Formulation In 10 mM PBS.
Gene Accession No. P63165
Gene Alias DAP-1/GMP1/OFC10/PIC1/SENP2/SMT3/SMT3C/SMT3H3/SUMO-1/UBL1
Gene Symbols SUMO1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human SUMO1.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7341
Test Specificity This antibody is specific to SUMO1 and shows no cross-reaction with either SUMO2 or SUMO3.
Target Species Human, Rat
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.