missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUMO2 Polyclonal Antibody
Rabbit Polyclonal Antibody
€ 618.00
Specifications
| Antigen | SUMO2 |
|---|---|
| Concentration | 0.20 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17179063
|
Invitrogen™
PA5111147 |
100 μL |
€ 618.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17159183
|
Invitrogen™
PA5111266 |
100 μL |
€ 618.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: TDKPAQLEME DEDTIDVFQQ PTGGV Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 92%.
SUMO2 is a member of the SUMO (small ubiquitin-like modifier) protein family. This protein family functions in a manner similar to ubiquitin in that it is bound to target proteins as part of a post-translational modification system. However, unlike ubiquitin which targets proteins for degradation, this protein is involved in a variety of cellular processes, such as nuclear transport, transcriptional regulation, apoptosis, and protein stability. In vertebrates, three members of the SUMO family have been described, SUMO 1 and the functionally distinct homologues SUMO 2 and SUMO 3. SUMO modification sites present in the N terminal regions of SUMO 2 and SUMO 3 are utilized by SAE1/SAE2 (SUMO E1) and Ubc9 (SUMO E2) to form polymeric chains of SUMO 2 and SUMO 3 on protein substrates, a property not shared by SUMO 1.Specifications
| SUMO2 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| SUMO2 | |
| HSMT3; sentrin 2; sentrin-2; small ubiquitin-like modifier 2; small ubiquitin-like modifier 2 L homeolog; small ubiquitin-like modifier 3; small ubiquitin-related modifier 2; Small ubiquitin-related modifier 2-A; small ubiquitin-related modifier 3; SMT3 homolog 1; SMT3 homolog 2; SMT3 suppressor of mif two 3 homolog 1; SMT3 suppressor of mif two 3 homolog 2; SMT3 suppressor of mif two 3 homolog 2 (yeast); SMT3 suppressor of mif two 3 homolog 3; SMT3 supressor of mif two 3 homolog 2; Smt3A; Smt3b; SMT3H1; SMT3H2; Sumo2; SUMO-2; sumo2.L; SUMO-2-A; sumo2-a; sumo2-b; SUMO3; SUMO-3; Ubiquitin-like protein SMT3A; ubiquitin-like protein SMT3B; XELAEV_18044053mg | |
| SUMO2 | |
| Primary | |
| Antigen affinity chromatography |
| 0.20 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P61956 | |
| 6613 | |
| Recombinant Protein corresonding to Human SUMO2. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title