missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUMO2/SUMO3 Recombinant Rabbit Monoclonal Antibody (6R8G7)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535631
This item is not returnable.
View return policy
Description
Immunogen sequence: MADEKPKEGV KTENNDHINL KVAGQDGSVV QFKIKRHTPL SKLMKAYCER QGLSMRQIRF RFDGQPINET DTPAQLEMED EDTIDVFQQQ TGGVY.
This gene encodes a protein that is a member of the SUMO (small ubiquitin-like modifier) protein family. It functions in a manner similar to ubiquitin in that it is bound to target proteins as part of a post-translational modification system. However, unlike ubiquitin which targets proteins for degradation, this protein is involved in a variety of cellular processes, such as nuclear transport, transcriptional regulation, apoptosis, and protein stability. It is not active until the last two amino acids of the carboxy-terminus have been cleaved off. Numerous pseudogenes have been reported for this gene. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
| SUMO2/SUMO3 | |
| Recombinant Monoclonal | |
| 3.0 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P55854, P61956, P61957, P61959, Q5XIF4, Q9Z172 | |
| SUMO2, Sumo3 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-95 of human SUMO2/3 (P61956). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 6R8G7 | |
| Unconjugated | |
| Sumo3 | |
| 2810014B19Rik; D10Ertd345e; GMP1; HSMT3; I79_000932; MIF2 suppressor; Sentrin; sentrin 2; sentrin-2; small ubiquitin-like modifier 2; small ubiquitin-like modifier 3; small ubiquitin-like modifier 3 L homeolog; small ubiquitin-related modifier 2; Small ubiquitin-related modifier 3; Smt3; SMT3 (supressor of mif two, 3) homolog 1; SMT3 homolog 1; SMT3 homolog 2; SMT3 homolog 3; SMT3 suppressor of mif two 3 homolog 1; SMT3 suppressor of mif two 3 homolog 2; SMT3 suppressor of mif two 3 homolog 2 (yeast); SMT3 suppressor of mif two 3 homolog 3; SMT3 suppressor of mif two 3 homolog 3 (yeast); SMT3 supressor of mif two 3 homolog 2; SMT3A; Smt3B; SMT3C; SMT3H1; SMT3H2; SMT3H3; SUMO-1; SUMO2; SUMO-2; Sumo3; SUMO-3; sumo3.L; Ubiquitin-like protein SMT3A; ubiquitin-like protein SMT3B; UBL1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 170930, 20610, 499417, 6612, 6613, 690244 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction