missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO4 Rabbit anti-Human, Mouse, Rat, Clone: 7X3V7, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16732-100UL
This item is not returnable.
View return policy
Description
SUMO4 Monoclonal antibody specifically detects SUMO4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SUMO4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 7X3V7 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| dJ281H8.4, IDDM5, small ubiquitin-like modifier 4 protein, Small ubiquitin-like protein 4, small ubiquitin-related modifier 4, SMT3 suppressor of mif two 3 homolog 2, SMT3 suppressor of mif two 3 homolog 4 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 4 (yeast), SMT3H4, SUMO-4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-95 of human SUMO4 (Q6EEV6). | |
| 100 μg | |
| Protein Phosphatase | |
| 387082 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction