missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation SunFire C18 OBD Prep Column
Click to view available options
Length (Metric):
50 mm
75 mm
100 mm
150 mm
Diameter (Metric):
19 mm
30 mm
Unit Size:
Each
Description
- The C18 ligand is highly retentive, especially for basic compounds and it is ideally suited for purification and impurity profile assays.
Specifications
Specifications
| Diameter (Metric) | 30 mm |
| Length (Metric) | 75 mm |
| Max. Pressure | 6000 psi (415 Bar) |
| Particle Shape | Spherical |
| Particle Size | 5 μm |
| Pore Size | 100 Å |
| Quantity | 1/Ea. |
| USP Type | L1 |
| Endcapped | Yes |
| Type | Preparative Column |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction