missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUOX Monoclonal Antibody (OTI5A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805951
This item is not returnable.
View return policy
Description
Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins.Specifications
| SUOX | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P51687 | |
| SUOX | |
| Human recombit protein fragment corresponding to amino acids 320-545 of human SUOX produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5A2 | |
| Unconjugated | |
| SUOX | |
| sulfite oxidase | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6821 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction