missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SUOX Recombinant Rabbit Monoclonal Antibody (JE56-74)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA544912

Product Code. 17849022

  • € 478.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SUOX Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow

Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SUOX
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
P51687, Q07116, Q8R086
SUOX
Recombinant protein within human Sulfite oxidase aa 100-200.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
JE56-74
Unconjugated
SUOX
SO; sulfite oxidase; sulfite oxidase, mitochondrial; Suox
Rabbit
Protein A
RUO
211389, 6821, 81805
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.