missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperFrost™ Microscope Slides, Clipped

The printed Thermo Scientific™ SuperFrost tab is impervious to virtually all common laboratory chemicals and reagents. A wide range of colors (7 standard colors) allows several examination specimens to be labeled and archived simply and clearly.

Brand:  Thermo Scientific™ AFAA000080##32E00MNZ10MH#

Additional Details : Weight : 2.50000kg

 View more versions of this product

Product Code. 12715318

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Time-consuming labeling with paper labels is no longer required
  • The writing area is resistant to all common solvents
  • SuperFrost slides may be sorted by color e.g. by patient, urgency, stain or examination method
  • Simple recognition of specified characteristics such as year of examination or department, use etc.
Specifications

Specifications

76mm
1mm
ISO8037/I
Clipped corners
50/Box, 5,000 in outer carton
26mm
White
Uncoated
90°, ground
Yes
Videos
Documents