missing translation for 'onlineSavingsMsg'
Learn More

SuperPicture™ Polymer Detection Kit, HRP, broad spectrum

Product Code. 10364213
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10364213

Brand: Invitrogen 878963

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

SuperPicture™ Polymer Detection Kit, HRP, broad spectrum

The SuperPicture™ Polymer Detection kit features our proprietary and enhanced HRP polymer technology to provide excellent sensitivity, high specificity, and a shorter
protocol than conventional immunohistochemical staining methods.

Our new polymer conjugation methods have resulted in a versatile polymer capable of intense nuclear, cytoplasmic, and membrane antigen staining. The polymer conjugate contains multiple HRP molecules to provide excellent sensitivity. The polymer staining method is faster and easier than a conventional LAB-SA protocol because it uses an HRP polymer conjugated to the second antibody and eliminates one incubation step. In addition, SuperPicture™ Polymer kits do not contain biotin nor streptavidin, so potential background due to endogenous biotin activity is completely avoided. Another advantage of SuperPicture™ Polymer detection system is the shorter polymer incubation time; the this system only requires 10 minutes, while leading competitors’ polymers require 30 minutes.

Reactivity: Mouse, Rabbit, Rat, G. Pig

Applications: Immunohistochemistry (FFPE)
TRUSTED_SUSTAINABILITY

Specifications

Description SuperPicture™ Polymer Detection Kit, Liquid, Soluble in Water, Wet Ice Shipping Condition, 2 to 8°C Storage Temperature, DAB, AEC Substrate, For Use With Light Microscope, For Clinical, Anatomical Pathology, Clinical Immunohistochemistry, Clinic
Product Type SuperPicture™ Polymer Detection Kit
Quantity 1 Kit
For Use With (Application) Anatomical Pathology, Clinical, Clinical Immunohistochemistry, Clinical IHC Staining and Detection
Detection Method Colorimetric
Form Liquid
Species Human
Content And Storage 2°C to 8°C
Packaging Quantity 1 Each

For In Vitro Diagnostic Use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.