Learn More
Memmert™ Support Basket With Perforated Stainless Steel Platform For Flask Clips, For Wnb22
Use these baskets with Memmert ovens. Memmert™ Support Baskets for Water Bath feature stainless steel perforated mounting shelves with grid perforations to take clips for conical flasks.
Brand:  Memmert™ W22B02782
Additional Details : Weight : 0.01100kg
Product Code. 16304657
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Includes: Stainless Steel perforated mounting shelf with grid perforations
Description
Description
  • Stainless steel support baskets
  • For Memmert Water Baths
Specifications
Specifications
14 L and 22 L Bath
Support Basket