Learn More
OHAUS™ Support Stand Rod
Shop All Ohaus Products
Click to view available options
Length (Metric):
460 mm
580 mm
710 mm
910 mm
1020 mm
1220 mm
1520 mm
Unit Size:
Each
Description
- Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
- Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
- Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
Specifications
Specifications
| Type | Rod |
| Material | Stainless Steel |
| Description | Clamp, Support, Rod 102cm, CLR-SPRODS102 |
| Length (Metric) | 1020 mm |