Learn More
OHAUS™ Support Stand Rod
Product Code. 15782449 Shop All Ohaus Products
Click to view available options
Length (Metric):
460 mm
580 mm
710 mm
910 mm
1020 mm
1220 mm
1520 mm
Unit Size:
Each
Product Code. 15782449
Brand: OHAUS™ 30400036
Only null left
Add to basket
Add to basket
OHAUS™ Support Stand Rod provides secure support for holding glassware at elevated positions.
  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
TRUSTED_SUSTAINABILITY
Specifications
Type Rod
Material Stainless Steel
Description Clamp, Support, Rod 46 cm, CLR-SPRODS046
Length (Metric) 460 mm