Learn More
OHAUS™ Support Stand Rod
Shop All Ohaus Products
Click to view available options
Length (Metric):
460 mm
580 mm
710 mm
910 mm
1020 mm
1220 mm
1520 mm
Förpackningsstorlek
Each
Beskrivning
- Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
- Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
- Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
Specifikationer
Specifikationer
| Type | Rod |
| Material | Stainless Steel |
| Description | Clamp, Support, Rod 46 cm, CLR-SPRODS046 |
| Length (Metric) | 460 mm |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering