missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Suppressor of Ty 4 homolog 1 Antibody (OTI6B10), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74412AF350
This item is not returnable.
View return policy
Description
Suppressor of Ty 4 homolog 1 Monoclonal specifically detects Suppressor of Ty 4 homolog 1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Suppressor of Ty 4 homolog 1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SUPT4H1 | |
| Full length recombinant protein of human Suppressor of Ty 4 homolog 1 (NP_003159) produced in E.coli. | |
| 0.1 mL | |
| 6827.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI6B10 | |
| Western Blot | |
| DRB sensitivity-inducing factor 14 kDa subunit, DRB sensitivity-inducing factor small subunit, DSIF p14, DSIF small subunit, SPT4, SPT4HhSPT4, suppressor of Ty (S.cerevisiae) 4 homolog 1, suppressor of Ty 4 homolog 1 (S. cerevisiae), SUPT4H, transcription elongation factor SPT4 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction