missing translation for 'onlineSavingsMsg'
Learn More

Suprofen, MedChemExpress

Product Code. 16474317
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474317 50 mg 50mg
16488438 10 mg 10mg
16448206 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16474317 Supplier Medchem Express Supplier No. HYB027050mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Suprofen (TN-762) is a non-steroidal anti-inflammatory drug (NSAID).

Suprofen (TN-762) is a non-steroidal anti-inflammatory drug (NSAID).

Specifications

Chemical Name or Material Suprofen
Physical Form Solid
CAS 40828-46-4
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C14H12O3S
For Use With (Application) COVID-19-immunoregulation
Synonym TN-762
Solubility Information DMSO : ≥ 100 mg/mL (384.16 mM)
SMILES O=C(O)C(C)C1=CC=C(C(C2=CC=CS2)=O)C=C1
Molecular Weight (g/mol) 260.31
Formula Weight 260.31
Percent Purity 99.11%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.