missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUPT16H Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-60942
This item is not returnable.
View return policy
Description
SUPT16H Polyclonal specifically detects SUPT16H in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| SUPT16H | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CDC68, Chromatin-specific transcription elongation factor 140 kDa subunit, facilitates chromatin remodeling 140 kDa subunit, Facilitates chromatin transcription complex subunit SPT16, FACT 140 kDa subunit, FACT complex subunit SPT16, FACT140, FACTp140, FACTP140FACT, FLJ10857, FLJ14010, FLJ34357, hSPT16, SPT16/CDC68, suppressor of Ty (S.cerevisiae) 16 homolog, suppressor of Ty 16 homolog (S. cerevisiae) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 11198 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1-0.3 ug/ml, Peptide ELISA Detection limit 1:8000 | |
| NP_009123.1 | |
| SUPT16H | |
| Peptide with sequence C-DLTNKEGKKPEEK corresponding to internal region according to NP_009123.1. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Pig, Mouse, Rat and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction