missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SUPT16H Polyclonal Antibody
GREENER_CHOICE

Product Code. 15779841 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15779841 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15779841 Supplier Invitrogen™ Supplier No. PA562045

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ASITSEVFNK FFKERVMEIV DADEKVRHSK LAESVEKAIE EKKYLAGADP STVEMCYPPI IQSGGNYNLK FSVVSDKNHM HFGAITCAMG IRFKSY Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Transcription of protein-coding genes can be reconstituted on naked DNA with only the general transcription factors and RNA polymerase II. However, this minimal system cannot transcribe DNA packaged into chromatin, indicating that accessory factors may facilitate access to DNA. One such factor, FACT, interacts specifically with histones H2A/H2B to effect nucleosome disassembly and transcription elongation. FACT is composed of an 80 kDa subunit and a 140 kDa subunit; this gene encodes the 140 kDa subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUPT16H
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SUPT16H
Gene Accession No. Q9Y5B9
Gene Alias Cdc68; Chromatin-specific transcription elongation factor 140 kDa subunit; chromatin-specific transcription elongation factor large subunit; DNA unwinding factor 140 kDa subunit; DUF140; facilitates chromatin remodeling 140 kDa subunit; facilitates chromatin transcription complex subunit spt16; FACT; FACT 140 kDa subunit; FACT complex subunit SPT16; FACT complex subunit SPT16-like protein; FACT140; Factp140; FLJ10857; FLJ14010; FLJ34357; fq44d08; HGNC:11465; hSPT16; SPT16; SPT16 homolog, facilitates chromatin remodeling subunit; SPT16/CDC68; suppressor of Ty 16; suppressor of Ty 16 homolog; suppressor of Ty 16 homolog (S. cerevisiae); Supt16; supt16h; wu:fq44d08; zgc:194708; zgc:194732
Gene Symbols SUPT16H
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SUPT16H. Recombinant protein control fragment (Product #RP-103053).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11198
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.