missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUPT4H1 Monoclonal Antibody (OTI5F12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803431S
This item is not returnable.
View return policy
Description
SUPT4H1 Monoclonal antibody specifically detects SUPT4H1 in Human samples. It is validated for Western BlotSpecifications
| SUPT4H1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P63272 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6827 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5F12 | |
| Unconjugated | |
| SUPT4H1 | |
| AL022777; DRB sensitivity-inducing factor 14 kDa subunit; DRB sensitivity-inducing factor small subunit; DRB sensitivity-inducing factor small subunit 1; DSIF p14; DSIF small subunit; DSIF small subunit 1; hSPT4; small hSpt4 subunit; SPT4; SPT4 homolog, DSIF elongation factor subunit; SPT4H; suppressor of Ty 4 homolog 1; suppressor of Ty 4 homolog 1 (S. cerevisiae); suppressor of Ty 4 homolog 2; suppressor of Ty 4A; Supt4a; Supt4h; Supt4h1; Supt4h1a; Supt4h2; transcription elongation factor SPT4; Transcription elongation factor SPT4 1; transcription elongation factor SPT4-A; zgc:92824; zgc:92824 protein | |
| Full length human recombinant protein of human SUPT4H1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction