missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUR1 Antibody (S289-16), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59320JF646
This item is not returnable.
View return policy
Description
SUR1 Monoclonal antibody specifically detects SUR1 in Human, Mouse, Rat, Hamster samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| SUR1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| ABC36, ATP-binding cassette, sub-family C (CFTR/MRP), member 8, HHF1Sulfonylurea receptor 1, HI, PHHIHRINS, sulfonylurea receptor (hyperinsulinemia), SUR1MRP8, SURATP-binding cassette transporter sub-family C member 8, TNDM2ATP-binding cassette sub-family C member 8 | |
| Fusion protein amino acids 1548-1582 (cytoplasmic C-terminus) of rat SUR1 | |
| 0.1 mL | |
| ABC Transporters, Lipid and Metabolism, Neuroscience | |
| 6833 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunofluorescence | |
| S289-16 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction