missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SURF5 Antibody (PCRP-MED22-2A7) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14191JF525
This item is not returnable.
View return policy
Description
SURF5 Monoclonal antibody specifically detects SURF5 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| SURF5 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Med24, mediator complex subunit 22SURF5surfeit 5, mediator of RNA polymerase II transcription subunit 22, MGC48682, Surf-5, Surfeit locus protein 5 | |
| Recombinant full-length human SURF5 protein (Uniprot: Q15528) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunoprecipitation, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-MED22-2A7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 6837 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction