missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Pursept™ A Xpress Surface Disinfecting Detergents

Disinfects in just 15 seconds. BRAND™ Pursept™ A Xpress Surface Disinfecting Detergents are effective against bacteria and fungi with little organic burden with mechanics. Limited virucidal efficacy including HIV, HBV, and HCV after 15 seconds.

Brand:  Brand™ 44830

Additional Details : CAS : 64-17-5 Weight : 9.18000kg Transport : UN number : 1170 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 11814110

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Includes: 1 atomizer head

Description

Description

Disinfects in just 15 seconds. Refillable pump-atomizer without gas. Pleasant, fresh fragrance.
  • DGHM/VAH tested and RKIcompliant, with aerosol-free application (disinfectant wipes).
  •  
Ingredients:
100 g Pursept™ A Xpress contain: 55 g ethanol, 0.03 g n-alkyl-aminopropyl-glycine, auxiliary agents.

Chemical and physical specifications:
  • Clear, colorless liquid,
  • Density: approx. 0.9 g/ml, pH value: approx. 7.5.
Microbiological efficiency of the undiluted working solution:
  • Surface disinfection: Bacteria and fungi with little organic burden with mechanics acc. to DGHM/VAH directives: 15 seconds.
  • Limited virucidal efficacy* (incl. HIV, HBV, HCV): 15 seconds.
  • Effectiveness against TbB: 30 seconds.
* As recommended by the RKI, Bundesgesundheitsblatt 01/2004.
Specifications

Specifications

Disinfectant Detergent
7.5
DGHM/VAH Tested and RKI Compliant
Bottle
10 Pk.
Liquid
Colorless
100g Pursept™ A Xpress Contain: 55g Ethanol, 0.03g n-Alkyl-Aminopropyl-Glycine, Auxiliary Agen
With Aerosol-Free Application
1 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • pursept A Xpress

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P271-Use only outdoors or in a well-ventilated area.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

Supplemental information
  • MIXTURE LIST-Contains : Ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers