Learn More
Surfactant Protein A Antibody (1G8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00653509-M03
This item is not returnable.
View return policy
Description
Surfactant Protein A Monoclonal antibody specifically detects Surfactant Protein A in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| Surfactant Protein A | |
| Monoclonal | |
| Unconjugated | |
| NP_001158118.1 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 653509 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 1G8 | |
| In 1x PBS, pH 7.4 | |
| COLEC4, PSAP, PSPA, PSP-A, SFTP1, SFTPA1B, SPA, SP-A, SPA1, SP-A1, surfactant protein A1B | |
| SFTPA1 (NP_001158118.1, 83 a.a. ∽ 199 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTVGEKVFSSNGQSITFDAIQEACARAGGRIAVPRNPEENEAIASFVKKYNTYAYVGLTEGPSPGDFRYSDGTPVNYTNWYRGEPAGRGKEQCVEMYTDGQWNDRNCLYSRLTICEF | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |