missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Surgical Masks
Click to view available options
:
Surgical Masks
Description
Order Info
dimProductType
Specifications
Specifications
| Product Type | Surgical Face Mask |
| Color | Light Blue |
| Certifications/Compliance | EN14683:2005 Type II |
| Filter Efficiency | High |
| For Use With (Application) | Designed to Optimal Protect the Patient |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction