missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Survivin Antibody (32.1), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-237MFV610
This item is not returnable.
View return policy
Description
Survivin Monoclonal antibody specifically detects Survivin in Human, Hamster samples. It is validated for Flow Cytometry
Specifications
| Survivin | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Hamster | |
| Purified |
| Flow Cytometry | |
| 32.1 | |
| Flow Cytometry | |
| API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha | |
| This Survivin Antibody (32.1) was developed against full length recombinant human Survivin [UniProt# O15392] | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells | |
| 332 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction