missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Survivin Antibody (60.11), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-238PE
This item is not returnable.
View return policy
Description
Survivin Monoclonal specifically detects Survivin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Knockdown Validated.
Specifications
| Survivin | |
| Monoclonal | |
| PE | |
| O15392 | |
| BIRC5 | |
| This Survivin Antibody (60.11) [PE] was developed against full length recombinant human Survivin [UniProt# O15392] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Survivin Antibody (60.11) [NB500-238] is specific for the cytoplasmic form of Survivin. | |
| Store at 4C in the dark. | |
| IgG2a κ |
| Flow Cytometry, KnockDown | |
| 60.11 | |
| Flow Cytometry, KnockDown Validated | |
| API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha | |
| Mouse | |
| 16 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells | |
| 332 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction