Learn More
Survivin Antibody (8E2), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB500-644APC
Product Code. 18226650
  • ONLINE EXCLUSIVE PRICE
    € 557.00
    € 526.05 / 0.10mL
    Save € 30.95
    5% Off
Description
Survivin Monoclonal specifically detects Survivin in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Survivin
Monoclonal
APC
O15392
BIRC5
This Survivin Antibody (8E2) [Allophycocyanin] was developed against full length recombinant human Survivin [UniProt# O15392]
Protein G purified
RUO
Primary
Human
Purified
Flow Cytometry
800
Flow Cytometry
API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha
Mouse
16 kDa
0.1 mL
Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells
332
Store at 4C in the dark.
IgG1 κ