missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Survivin Antibody (91630) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB886M
This item is not returnable.
View return policy
Description
Survivin Monoclonal antibody specifically detects Survivin in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| Survivin | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 91630 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha | |
| E. coli-derived recombinant human Survivin, Met1-Asp142, Accession # O15392 | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Stem Cells | |
| 332 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction