missing translation for 'onlineSavingsMsg'
Learn More

Survivin Antibody (BIRC5/8936R), Novus Biologicals™

Product Code. 18691844 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18691844 20 μg 20µg
18672425 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18691844 Supplier Novus Biologicals Supplier No. NBP32379020ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Survivin Monoclonal antibody specifically detects Survivin in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Survivin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BIRC5/8936R
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Alias API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha
Host Species Rabbit
Immunogen Recombinant fragment (around aa1-100) of human Survivin protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 332
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.