Learn More
SUSD1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-61129MFV450
Product Code. 18477447
  • € 588.00 / 0.10mL
Description
SUSD1 Polyclonal antibody specifically detects SUSD1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SUSD1
mFluor Violet 450 SE
50mM Sodium Borate
0.1 mL
Western Blot, ELISA
Western Blot, ELISA
Rabbit
Cancer, Cell Cycle and Replication