missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUSD2 Antibody (1279A) [DyLight 680], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB90562Y
This item is not returnable.
View return policy
Description
SUSD2 Monoclonal antibody specifically detects SUSD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| SUSD2 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| 1279A | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2 | |
| HEK293 human embryonic kidney cell line-derived human SUSD2, Met1-Ala785, Accession # Q9UGT4 | |
| 0.1 mL | |
| Cancer | |
| 56241 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction