missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUSD2 Antibody (1279B), DyLight 405, R&D Systems™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals MAB90564V
This item is not returnable.
View return policy
Description
SUSD2 Monoclonal specifically detects SUSD2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| SUSD2 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SUSD2 | |
| Partial recombinant human SUSD2 protein (amino acids 1-785) [UniProt Q9UGT4] | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| 1279B | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 56241 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction