missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUSD2 Antibody (1279C), DyLight 680, R&D Systems™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals MAB90565FR
This item is not returnable.
View return policy
Description
SUSD2 Monoclonal specifically detects SUSD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| SUSD2 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SUSD2 | |
| Partial recombinant human SUSD2 protein (amino acids 1-785) [UniProt Q9UGT4] | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| 1279C | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 56241 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction