missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUSD2 Antibody (944827) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB90561F
This item is not returnable.
View return policy
Description
SUSD2 Monoclonal antibody specifically detects SUSD2 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| SUSD2 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 944827 | |
| Flow Cytometry, CyTOF-ready | |
| BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2 | |
| Human embryonic kidney cell line HEK293-derived human SUSD2, Met1-Ala785, Accession # Q9UGT4 | |
| 0.1 mL | |
| Cancer | |
| 56241 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction