missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUSD3 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82017AF594
This item is not returnable.
View return policy
Description
SUSD3 Polyclonal antibody specifically detects SUSD3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| SUSD3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 594 | |
| MGC26847, sushi domain containing 3, sushi domain-containing protein 3 | |
| SUSD3 antibody was raised against a 16 amino acid peptide from near the center of human SUSD3. The immunogen is located within amino acids 120 - 170 of SUSD3. Amino Acid Squence: VKKSKRRRSNRSAQL | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 203328 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction