Learn More
B Braun™ Suture Clips
Description
- Manufactured from stainless steel
- 7.5 x 1.75 mm
- Provided in packs of 100
Specifications
Specifications
| Product Type | Suture Clip |
| Length (Metric) | 7.5 mm |
| Material | Stainless Steel |
| Dimensions (L x W) | 7.5 x 1.75 mm |