missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUV3L1 Antibody (244), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43565
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry (Paraffin)
SUV3L1 Monoclonal specifically detects SUV3L1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SUV3L1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| SUPV3L1 | |
| The immunogen used to generate this antibody corresponds to human SUPV3L1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 244 | |
| Western Blot 1:500-1:10000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| ATP-dependent RNA helicase SUPV3L1, mitochondrial, EC 3.6.1, EC 3.6.4.13, suppressor of var1 (S.cerevisiae) 3-like 1, Suppressor of var1 3-like protein 1, suppressor of var1, 3-like 1 (S. cerevisiae), SUV3-like protein 1, SUV3suppressor of var1, 3-like 1(SUV3) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6832 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction