missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUV420h1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 213.00 - € 450.00
Specifications
| Antigen | SUV420h1 |
|---|---|
| Concentration | 1.16 mg/mL |
| Dilution | Flow Cytometry 2-5 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200, Western Blot (Negative) |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18096466
|
Novus Biologicals
NBP1-97313SS |
0.025 mL |
€ 213.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18088716
|
Novus Biologicals
NBP1-97313 |
0.1 mL |
€ 450.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SUV420h1 Polyclonal specifically detects SUV420h1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative).Specifications
| SUV420h1 | |
| Flow Cytometry 2-5 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200, Western Blot (Negative) | |
| Polyclonal | |
| Rabbit | |
| Cancer, Epigenetics, Transcription Factors and Regulators | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| C630029K18Rik, CGI85, CGI-85, EC 2.1.1.43, histone-lysine N-methyltransferase SUV420H1, KMT5B, KMT5B Lysine N-methyltransferase 5B, MGC118906, MGC118909, MGC21161, MGC703, Su(var)4-20 homolog 1, Suppressor of variegation 4-20 homolog 1, suppressor of variegation 4-20 homolog 1 (Drosophila), suv4-20h1 | |
| KMT5B | |
| IgG | |
| Affinity Purified |
| 1.16 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q4FZB7, Q3U8K7 | |
| 51111 | |
| A synthetic peptide made to an internal portion of the human SUV420h1 protein (between residues 450-500) [UniProt Q4FZB7] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title