missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUV420H2 Antibody (RAB-C409), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20151CL1
This item is not returnable.
View return policy
Description
SUV420H2 Monoclonal antibody specifically detects SUV420H2 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Mass Spectrometry,ChIP assay (ChIP),Immunocytochemistry/Immunofluorescence
Specifications
| SUV420H2 | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 2.1.1.43, FLJ98627, histone-lysine N-methyltransferase SUV420H2, KMT5Csuv4-20h2, Lysine N-methyltransferase 5C, MGC2705, Su(var)4-20 homolog 2, Suppressor of variegation 4-20 homolog 2, suppressor of variegation 4-20 homolog 2 (Drosophila), Suv4-20h2 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing SUV420H2 protein under non-denaturing conditions. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunoprecipitation, Mass Spectrometry, ChIP Assay | |
| RAB-C409 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 84787 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction