missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUZ12 Antibody (2B9G12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61892-0.025ml
This item is not returnable.
View return policy
Description
SUZ12 Monoclonal specifically detects SUZ12 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| SUZ12 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| ChET 9 protein, CHET9polycomb protein SUZ12, Chromatin precipitated E2F target 9 protein, JJAZ1Joined to JAZF1 protein, KIAA0160Suppressor of zeste 12 protein homolog, suppressor of zeste 12 homolog (Drosophila) | |
| Mouse | |
| 83 kDa | |
| 0.025 mL | |
| 23512 | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 2B9G12 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| SUZ12 | |
| Purified recombinant fragment of human SUZ12 (AA: 1-139) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction