missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SV2A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29314
This item is not returnable.
View return policy
Description
SV2A Polyclonal antibody specifically detects SV2A in Human samples. It is validated for Immunoprecipitation
Specifications
| SV2A | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9900 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| KIAA0736SV2, synaptic vesicle glycoprotein 2A | |
| Between 1 and 50 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction