missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SV2B Monoclonal Antibody (2C6)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524430
This item is not returnable.
View return policy
Description
Peptide Sequence: YFQDEEYKSK MKVFFGEHVY GATINFTMEN QIHQHGKLVN DKFTRMYFKH VLFEDTFFDE.
Probably plays a role in the control of regulated secretion in neural and endocrine cells.
Specifications
| SV2B | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q7L1I2 | |
| Sv2b | |
| SV2B (NP_055663, 417 a.a. ~ 476 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA | |
| 2C6 | |
| Unconjugated | |
| Sv2b | |
| A830038F04Rik; AI850768; HsT19680; Kiaa0735; mKIAA0735; SV2B; Sv2bb; synaptic vesicle glycoprotein 2 b; synaptic vesicle glycoprotein 2b; Synaptic vesicle protein 2B; synaptic vesicle protein 2B homolog | |
| Mouse | |
| Protein A | |
| RUO | |
| 9899 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction